Oil Red O solution
Synonym(s):
Solvent Red 27, Sudan Red 5B
Empirical Formula (Hill Notation):
C26H24N4O
CAS Number:
Molecular Weight:
408.49
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing