Notoginsenoside R1
Synonym(s):
(3β,6α,12β)-20-(β-D-Glucopyranosyloxy)-3,12-dihydroxydammar-24-en-6-yl 2-O-β-D-xylopyranosyl-β-D-glucopyranoside, Sanchinoside R1
Empirical Formula (Hill Notation):
C47H80O18
CAS Number:
Molecular Weight:
933.13
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing