Norfloxacin Related Compound E
Synonym(s):
7-Chloro-1-ethyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid
Empirical Formula (Hill Notation):
C16H18ClN3O3
CAS Number:
Molecular Weight:
335.79
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing