Skip to Content
Merck
N-Nitrosoethylisopropylamine analytical standard

N-Nitroso-ethyl-isopropylamine

N-Nitrosoethylisopropylamine analytical standard
Synonym(s):
N-Ethyl-N-nitroso-2-propanamine solution, N-Ethyl-N-nitroso-2-propanamine, EIPNA, NEIPA
Empirical Formula (Hill Notation):
C5H12N2O
CAS Number:
Molecular Weight:
116.16

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique