Skip to Content
Merck
vibrant-m

N-Nitrosodipropylamine

vibrant-m
Synonym(s):
N-Nitrosodi-n-propylamine, NDPA
Empirical Formula (Hill Notation):
C6H14N2O
CAS Number:
Molecular Weight:
130.19

Products

Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique