Skip to Content
Merck
N,N′-Disuccinimidyl oxalate 85%

N,N′-Disuccinimidyl oxalate

N,N′-Disuccinimidyl oxalate 85%
Synonym(s):
Di(N-succinimidyl) oxalate, N,N′-Disuccinimidyl oxalate, DSO
Empirical Formula (Hill Notation):
C10H8N2O8
CAS Number:
Molecular Weight:
284.18

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing