Skip to Content
Merck
N-Methyl-1-naphthalenemethylamine hydrochloride 98%

N-Methyl-1-naphthalenemethylamine hydrochloride

N-Methyl-1-naphthalenemethylamine hydrochloride 98%
Linear Formula:
C10H7CH2NHCH3 · HCl
CAS Number:
Molecular Weight:
207.70

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing