![Nicotine Related Compound B pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/258/751/999d5d43-e07f-4e0d-a3bd-eeff9aba35ea/640/999d5d43-e07f-4e0d-a3bd-eeff9aba35ea.jpg)
Nicotine Related Compound B
![Nicotine Related Compound B pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/images/258/751/999d5d43-e07f-4e0d-a3bd-eeff9aba35ea/640/999d5d43-e07f-4e0d-a3bd-eeff9aba35ea.jpg)
Synonym(s):
β-Nicotyrine, 3-(1-Methyl-1H-pyrrol-2-yl)pyridine
Empirical Formula (Hill Notation):
C10H10N2
CAS Number:
Molecular Weight:
158.20
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2