Skip to Content
Merck
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride BioXtra

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride BioXtra
Synonym(s):
N-Ethyl-N′-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDAC, EDC, EDC hydrochloride, WSC hydrochloride
Empirical Formula (Hill Notation):
C8H17N3 · HCl
CAS Number:
Molecular Weight:
191.70
EC Number:

Products

Brand
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Reaction Type
Reagent Type
Special Grade
Technique