Skip to Content
Merck
Mycophenolic acid analytical standard

Mycophenolic acid

Mycophenolic acid analytical standard
Synonym(s):
6-(1,3-Dihydro-7-hydroxy-5-methoxy-4-methyl-1-oxoisobenzofuran-6-yl)-4-methyl-4-hexanoic acid, 6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-5-phthalanyl)-4-methyl-4-hexenoic acid, NSC 129185, Mycophenolic Acid
Empirical Formula (Hill Notation):
C17H20O6
CAS Number:
Molecular Weight:
320.34
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Technique