![Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 (KDR) antibody produced in mouse clone 260.4, KDR-2, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/321/359/23688fa0-ca4e-4d4c-920e-d784931c040a/640/23688fa0-ca4e-4d4c-920e-d784931c040a.jpg)
Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 (KDR) antibody produced in mouse
![Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 (KDR) antibody produced in mouse clone 260.4, KDR-2, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/321/359/23688fa0-ca4e-4d4c-920e-d784931c040a/640/23688fa0-ca4e-4d4c-920e-d784931c040a.jpg)
Synonym(s):
Monoclonal Anti-Vascular Endothelial Growth Factor Receptor-2 antibody produced in mouse, Anti-KDR, Anti-VEGFR-2
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing