
Monoclonal Anti-β-COP antibody produced in mouse

Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2