Skip to Content
Merck
vibrant-m

Metoclopramide impurity A

vibrant-m
Synonym(s):
4-(Acetylamino)-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide
Empirical Formula (Hill Notation):
C16H24ClN3O3
CAS Number:
Molecular Weight:
341.83

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing