![Methyl green zinc chloride double salt (C.I. 42590)](/deepweb/assets/sigmaaldrich/product/images/312/035/4a957182-0d55-46aa-b928-fd7a4333514f/640/4a957182-0d55-46aa-b928-fd7a4333514f.jpg)
Methyl green zinc chloride double salt (C.I. 42590)
![Methyl green zinc chloride double salt (C.I. 42590)](/deepweb/assets/sigmaaldrich/product/images/312/035/4a957182-0d55-46aa-b928-fd7a4333514f/640/4a957182-0d55-46aa-b928-fd7a4333514f.jpg)
Empirical Formula (Hill Notation):
C27H35BrClN3 · ZnCl2
CAS Number:
Molecular Weight:
653.24
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing