![Massoia Lactone natural, ≥95%, FG](/deepweb/assets/sigmaaldrich/product/structures/148/369/0ad67575-d7c8-48fc-b555-ce4bbe9014b5/640/0ad67575-d7c8-48fc-b555-ce4bbe9014b5.png)
Massoia Lactone
![Massoia Lactone natural, ≥95%, FG](/deepweb/assets/sigmaaldrich/product/structures/148/369/0ad67575-d7c8-48fc-b555-ce4bbe9014b5/640/0ad67575-d7c8-48fc-b555-ce4bbe9014b5.png)
Synonym(s):
6-Pentyl-5,6-dihydropyran-2-one
Empirical Formula (Hill Notation):
C10H16O2
CAS Number:
Molecular Weight:
168.23
Products
Product Category
Brand
Sustainability
Biological Source
Feature
Formula Weight
Purity
Agency
Tissue
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing