Skip to Content
Merck
Lurasidone hydrochloride ≥98% (HPLC)

Lurasidone hydrochloride

Lurasidone hydrochloride ≥98% (HPLC)
Synonym(s):
Lurasidone hydrochloride solution
Empirical Formula (Hill Notation):
C28H36N4O2S · HCl
CAS Number:
Molecular Weight:
529.14

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing