Lofepramine hydrochloride
Synonym(s):
N-(4-Chlorobenzoylmethyl)-3-(10,11-dihydro-5h-dibenz[b,f]azepin-5-yl)-n-methylpropylamine hydrochloride
Empirical Formula (Hill Notation):
C26H28Cl2N2O
CAS Number:
Molecular Weight:
455.42
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing