Lenacil
Synonym(s):
3-Cyclohexyl-6,7-dihydro-1H-cyclopentapyrimidine-2,4-(3H,5H)-dione
Empirical Formula (Hill Notation):
C13H18N2O2
CAS Number:
Molecular Weight:
234.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2