Skip to Content
Merck
Lead(II) nitrate ≥99.95% trace metals basis

Lead(II) nitrate

Lead(II) nitrate ≥99.95% trace metals basis
Synonym(s):
Lead dinitrate, Lead(II) nitrate
Linear Formula:
Pb(NO3)2
CAS Number:
Molecular Weight:
331.21
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade