Skip to Content
Merck
Lansoprazole Related Compound A pharmaceutical secondary standard, certified reference material

Lansoprazole Related Compound A

Lansoprazole Related Compound A pharmaceutical secondary standard, certified reference material
Synonym(s):
2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfonyl]-1H-benzimidazole
Empirical Formula (Hill Notation):
C16H14F3N3O3S
CAS Number:
Molecular Weight:
385.36

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade