Skip to Content
Merck
Lactose octaacetate 95%

Lactose octaacetate

Lactose octaacetate 95%
Empirical Formula (Hill Notation):
C28H38O19
CAS Number:
Molecular Weight:
678.59

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing