![KiCqStart® Probe qPCR ReadyMix™, Low ROX™ with Low ROX for ABI and Stratagene instruments](/deepweb/assets/sigmaaldrich/product/images/131/864/a2681a6d-a721-47ae-bbdb-d8e0e15228af/640/a2681a6d-a721-47ae-bbdb-d8e0e15228af.jpg)
KiCqStart® Probe qPCR ReadyMix™, Low ROX™
![KiCqStart® Probe qPCR ReadyMix™, Low ROX™ with Low ROX for ABI and Stratagene instruments](/deepweb/assets/sigmaaldrich/product/images/131/864/a2681a6d-a721-47ae-bbdb-d8e0e15228af/640/a2681a6d-a721-47ae-bbdb-d8e0e15228af.jpg)
Synonym(s):
Kicqstart® Probe qPCR ReadyMix™
Products
Product Category
Brand
Color
Physical Form
Number of Reactions
dNTPs
Hot Start
Sample Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing