
Isoalantolactone

Synonym(s):
Isohelenin
Empirical Formula (Hill Notation):
C15H20O2
CAS Number:
Molecular Weight:
232.32
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing