![Horse serum liquid](/deepweb/assets/sigmaaldrich/product/images/361/678/09c08fe0-9f58-47eb-b7a7-dffa7f199290/640/09c08fe0-9f58-47eb-b7a7-dffa7f199290.jpg)
Horse Serum
![Horse serum liquid](/deepweb/assets/sigmaaldrich/product/images/361/678/09c08fe0-9f58-47eb-b7a7-dffa7f199290/640/09c08fe0-9f58-47eb-b7a7-dffa7f199290.jpg)
Synonym(s):
HS, equine sera, equine serum, horse sera, sera, serum, Serum
Products
Product Category
Brand
Biological Source
Physical Form
Tissue
Chemical Composition
Quality Segment
Geographic Origin
Sterility
Sterilization Method
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing