Homoharringtonine
Synonym(s):
Ceflatonin, Cephalotaxine 4-methyl (2R)-2-hydroxy-2-(4-hydroxy-4-methylpentyl)butanedioate, Myelostat, NSC 141633, Omacetaxine mepesuccinate
Empirical Formula (Hill Notation):
C29H39NO9
CAS Number:
Molecular Weight:
545.62
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing