Hibifolin
Synonym(s):
3,3′,4′,5,7,8-Hexahydroxyflavone 8-glucuronide, Gossypetin 8-glucuronide
Empirical Formula (Hill Notation):
C21H18O14
CAS Number:
Molecular Weight:
494.36
Products
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing