![HEPES sodium salt ≥99.5% (titration), free-flowing, Redi-Dri™](/deepweb/assets/sigmaaldrich/product/structures/418/922/eef66454-9725-4b04-a17b-6080d47fde92/640/eef66454-9725-4b04-a17b-6080d47fde92.png)
HEPES sodium salt
![HEPES sodium salt ≥99.5% (titration), free-flowing, Redi-Dri™](/deepweb/assets/sigmaaldrich/product/structures/418/922/eef66454-9725-4b04-a17b-6080d47fde92/640/eef66454-9725-4b04-a17b-6080d47fde92.png)
Synonym(s):
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt, HEPES, Sodium Salt, ULTROL® Grade
Empirical Formula (Hill Notation):
C8H17N2NaO4S
CAS Number:
Molecular Weight:
260.29
Products
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
pH Value
Physical Form
pK Value
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing