Skip to Content
Merck
Guaiacol Impurity H Pharmaceutical Secondary Standard; Certified Reference Material

Guaiacol Impurity H

Guaiacol Impurity H Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
3-Methoxyphenol, 3-Hydroxyanisol, Resorcinol monomethyl ether
Linear Formula:
CH3OC6H4OH
CAS Number:
Molecular Weight:
124.14

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1