
Gonyautoxin-1 and Gonyautoxin-4 solution

Synonym(s):
GTX-1 - GTX-4 mixture
Empirical Formula (Hill Notation):
C10H17N7O9S
Molecular Weight:
411.35
Products
Brand
Formula Weight
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing