Skip to Content
Merck
vibrant-m

Glipizide Related Compound C

vibrant-m
Synonym(s):
1-cyclohexyl-3-[4-[2-[[(-methylpyrazin-2-yl)carnonyl]amino]ethyl]phenyl]sulfonyl]ureea (Glipizide Related Compound C-USP
Empirical Formula (Hill Notation):
C21H27N5O4S
Molecular Weight:
445.54

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1