![Fosfomycin sodium VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/386/960/03f38d0b-ae06-4352-b010-aea00c9c2f69/640/03f38d0b-ae06-4352-b010-aea00c9c2f69.png)
Fosfomycin sodium
![Fosfomycin sodium VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/386/960/03f38d0b-ae06-4352-b010-aea00c9c2f69/640/03f38d0b-ae06-4352-b010-aea00c9c2f69.png)
Synonym(s):
Phosphomycin disodium salt, (−)-(1R,2S)-(1,2-Epoxypropyl)phosphonic acid, Fosfomycin, Phosphonomycin
Empirical Formula (Hill Notation):
C3H5Na2O4P
Molecular Weight:
182.02
EC Number:
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing