![Fosamprenavir calcium United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/275/216/8a1864fa-64f7-4ece-a611-2864c43a59e2/640/8a1864fa-64f7-4ece-a611-2864c43a59e2.png)
Fosamprenavir calcium
![Fosamprenavir calcium United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/275/216/8a1864fa-64f7-4ece-a611-2864c43a59e2/640/8a1864fa-64f7-4ece-a611-2864c43a59e2.png)
Synonym(s):
Calcium (3S) tetrahydro-3-furanyl(1S,2R)-3-[[(4-aminophenyl)sulfonyl](isobutyl)amino]-1 -benzyl-2-(phosphonooxy)propylcarbamate, GW433908
Empirical Formula (Hill Notation):
C25H34N3O9PS · Ca
CAS Number:
Molecular Weight:
623.67
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing