Skip to Content
Merck
Fmoc-Lys(Boc)-OH ≥98.0% (HPLC)

Fmoc-Lys(Boc)-OH

Fmoc-Lys(Boc)-OH ≥98.0% (HPLC)
Synonym(s):
Nα-Fmoc-Nε-Boc-L-lysine, Fmoc-Lys(Boc)-OH
Empirical Formula (Hill Notation):
C26H32N2O6
CAS Number:
Molecular Weight:
468.54

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing