![Fmoc-Leu-OH ≥97.0%](/deepweb/assets/sigmaaldrich/product/structures/169/131/fdeb0034-afb1-476d-a96c-dad69dc957db/640/fdeb0034-afb1-476d-a96c-dad69dc957db.png)
Fmoc-Leu-OH
![Fmoc-Leu-OH ≥97.0%](/deepweb/assets/sigmaaldrich/product/structures/169/131/fdeb0034-afb1-476d-a96c-dad69dc957db/640/fdeb0034-afb1-476d-a96c-dad69dc957db.png)
Synonym(s):
N-(9-Fluorenylmethoxycarbonyl)-L-leucine, Fmoc-L-leucine, Fmoc-Leu-OH
Empirical Formula (Hill Notation):
C21H23NO4
CAS Number:
Molecular Weight:
353.41
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing