Skip to Content
Merck
vibrant-m

Flumazenil Related Compound B

vibrant-m
Synonym(s):
Ethyl 8-hydroxy-5-methyl-6-oxo-5,6-dihydro-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
Empirical Formula (Hill Notation):
C15H15N3O4
CAS Number:
Molecular Weight:
301.30

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1