Skip to Content
Merck
Ethyl 2-furoate ≥98%, FG

Ethyl 2-furoate

Ethyl 2-furoate ≥98%, FG
Synonym(s):
Ethyl furan-2-carboxylate
Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing