Ethionamide for system suitability
Synonym(s):
Ethionamide, 2-Ethyl-4-pyridinecarbothioamide
Empirical Formula (Hill Notation):
C8H10N2S
CAS Number:
Molecular Weight:
166.24
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing