Skip to Content
Merck
Ethanethiol ≥97%

Ethanethiol

Ethanethiol ≥97%
Synonym(s):
Ethyl mercaptan, Mercaptan C2, Ethanethiol
Linear Formula:
C2H5SH
CAS Number:
Molecular Weight:
62.13
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing