![Eriocitrin primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/102/928/115261d1-5107-41f1-9ce9-b0b05612bb41/640/115261d1-5107-41f1-9ce9-b0b05612bb41.png)
Eriocitrin
![Eriocitrin primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/102/928/115261d1-5107-41f1-9ce9-b0b05612bb41/640/115261d1-5107-41f1-9ce9-b0b05612bb41.png)
Synonym(s):
(S)-3′,4′,5,7-Tetrahydroxyflavanone-7-[6-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], Eriodictioside, Eriodictyol 7-O-rutinoside
Empirical Formula (Hill Notation):
C27H32O15
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing