Skip to Content
Merck
Endosulfan sulfate certified reference material

Endosulfan sulfate

Endosulfan sulfate certified reference material
Synonym(s):
Endosulfan sulfate
Empirical Formula (Hill Notation):
C9H6Cl6O4S
CAS Number:
Molecular Weight:
422.92

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing