![Dorzolamide hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/414/336/8b3b2fa2-b01a-4e2c-a4fb-c348dab85528/640/8b3b2fa2-b01a-4e2c-a4fb-c348dab85528.png)
Dorzolamide hydrochloride
![Dorzolamide hydrochloride British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/414/336/8b3b2fa2-b01a-4e2c-a4fb-c348dab85528/640/8b3b2fa2-b01a-4e2c-a4fb-c348dab85528.png)
Synonym(s):
Dorzolamide hydrochloride
Empirical Formula (Hill Notation):
C10H16N2O4S3 · HCl
CAS Number:
Molecular Weight:
360.90
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing