Skip to Content
Merck
vibrant-m

Domperidone maleate

vibrant-m
Empirical Formula (Hill Notation):
C22H24ClN5O2 · C4H4O4
CAS Number:
Molecular Weight:
541.98

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade