Skip to Content
Merck
(±)-AMPA hydrobromide ≥98% (HPLC)

(±)-AMPA hydrobromide

(±)-AMPA hydrobromide ≥98% (HPLC)
Synonym(s):
(±)-α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid hydrobromide
Empirical Formula (Hill Notation):
C7H10N2O4 · HBr
CAS Number:
Molecular Weight:
267.08

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing