Skip to Content
Merck
DL-4-Hydroxy-3-methoxymandelic acid ≥95%, FG

DL-4-Hydroxy-3-methoxymandelic acid

DL-4-Hydroxy-3-methoxymandelic acid ≥95%, FG
Synonym(s):
α,4-Dihydroxy-3-methoxybenzeneacetic acid, VMA, Vanillomandelic acid, Vanillylmandelic acid, Vanilmandelic acid
Linear Formula:
HOC6H3(OCH3)CH(OH)CO2H
CAS Number:
Molecular Weight:
198.17

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Organoleptic Class
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing