![DL-3-(4-Hydroxyphenyl)lactic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/696/255/4e184673-8d6c-46f1-85ec-d72ed133db26/640/4e184673-8d6c-46f1-85ec-d72ed133db26.png)
DL-3-(4-Hydroxyphenyl)lactic acid
![DL-3-(4-Hydroxyphenyl)lactic acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/696/255/4e184673-8d6c-46f1-85ec-d72ed133db26/640/4e184673-8d6c-46f1-85ec-d72ed133db26.png)
Synonym(s):
DL-p-Hydroxyphenyllactic acid, 3-(4-Hydroxyphenyl)-2-hydroxypropanoic acid
Empirical Formula (Hill Notation):
C9H10O4
CAS Number:
Molecular Weight:
182.17
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing