Skip to Content
Merck
Dimethyl succinate analytical standard

Dimethyl succinate

Dimethyl succinate analytical standard
Synonym(s):
Succinic acid dimethyl ester, Dimethyl succinate, Dimethylsuccinate
Linear Formula:
CH3OCOCH2CH2COOCH3
CAS Number:
Molecular Weight:
146.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing