Skip to Content
Merck
δ-Damascone analytical standard

δ-Damascone

δ-Damascone analytical standard
Synonym(s):
1-(2,6,6-Trimethyl-3-cyclohexen-1-yl)-2-buten-1-one
Empirical Formula (Hill Notation):
C13H20O
CAS Number:
Molecular Weight:
192.30
EC Number:

Products

Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing