Skip to Content
Merck
(+)-Abscisic acid analytical standard

(+)-Abscisic acid

(+)-Abscisic acid analytical standard
Synonym(s):
(S)-5-(1-Hydroxy-2,6,6-trimethyl-4-oxo-2-cyclohexen-1-yl)-3-methyl-(2Z,4E)-pentadienoic acid, ABA, Dormin, (+)-Abscisic Acid
Empirical Formula (Hill Notation):
C15H20O4
CAS Number:
Molecular Weight:
264.32
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade