Skip to Content
Merck
Cyclophosphamide British Pharmacopoeia (BP) Reference Standard

Cyclophosphamide

Cyclophosphamide British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Cyclophosphamide monohydrate, 2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine 2-oxide, Cytoxan
Empirical Formula (Hill Notation):
C7H15Cl2N2O2P · H2O
CAS Number:
Molecular Weight:
279.10

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique