![Cyclomethicone 4 United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/208/960/1eb9c41e-76bc-4115-9833-eb813fb71935/640/1eb9c41e-76bc-4115-9833-eb813fb71935.png)
Cyclomethicone 4
![Cyclomethicone 4 United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/208/960/1eb9c41e-76bc-4115-9833-eb813fb71935/640/1eb9c41e-76bc-4115-9833-eb813fb71935.png)
Synonym(s):
Octamethylcyclotetrasiloxane
Linear Formula:
[-Si(CH3)2O-]4
CAS Number:
Molecular Weight:
296.62
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing